Learn More
Medchemexpress LLC Camptothecin-20S-O-p 50mg | 194414-69-2 | 404.42 g·mol⁻1 | C23H20N2O5 | 50 MG
SDP
Catalog No. 5000308183
Encompass_Preferred
Camptothecin-20(S)-O-propionate (CZ48) is the C20-propionate ester prodrug of camptothecin that functions as a topoisomerase I inhibitor and is used in anticancer research as a small-molecule investigational compound.
  • Acts as a topoisomerase I inhibitor.
  • Is the C20-propionate ester (prodrug) of camptothecin.
  • Supplied at research quantities suitable for in vitro and in vivo studies.
  • High purity as verified by certificate of analysis (99.55%).
  • Chemical formula C23H20N2O5; molecular weight 404.42 g·mol⁻1.
  • Accompanied by a data sheet and certificate of analysis.
Catalog No. 50-003-08183 Supplier Medchemexpress LLC Supplier No. HY11466850MG
Camptothecin-20(S)-O-propionate (CZ48) is the C20-propionate ester prodrug of camptothecin that functions as a topoisomerase I inhibitor and is used in anticancer research as a small-molecule investigational compound.
  • Acts as a topoisomerase I inhibitor.
  • Is the C20-propionate ester (prodrug) of camptothecin.
  • Supplied at research quantities suitable for in vitro and in vivo studies.
  • High purity as verified by certificate of analysis (99.55%).
  • Chemical formula C23H20N2O5; molecular weight 404.42 g·mol⁻1.
  • Accompanied by a data sheet and certificate of analysis.