Learn More
Apexbio Technology LLC Camptothecin(Synonyms: CPT, Camptothecine, (+)-Camptothecin, NSC-94600, (S)-(+)-Camptothecin, Topotecin, 20(S)-Camptothecin), 250mg, CAS: 2114454.
SDP
Catalog No. 5000060770
Encompass_Preferred
Camptothecin (CAS 7689-03-4) is a small molecule inhibitor that selectively targets topoisomerase I exhibiting an IC50 value of 679 nM It causes DNA damage evident by phosphorylation of histone H2AX ( H2AX) leading to tumor cell death and cellular senescence via the ATM-Chk2-p53-p21 signaling pathway Additionally camptothecin induces autophagy through activation of the AMPK-TSC2-mTOR pathway Preclinical studies demonstrate its anti-tumor activity against tumor cell lines such as SMMC-7721 MCF-7 HCT-116 and NSCLC xenografts underscoring its utility in cancer biology research
Catalog No. 50-000-60770 Supplier Apexbio Technology LLC Supplier No. A2877250
Camptothecin (CAS 7689-03-4) is a small molecule inhibitor that selectively targets topoisomerase I exhibiting an IC50 value of 679 nM It causes DNA damage evident by phosphorylation of histone H2AX ( H2AX) leading to tumor cell death and cellular senescence via the ATM-Chk2-p53-p21 signaling pathway Additionally camptothecin induces autophagy through activation of the AMPK-TSC2-mTOR pathway Preclinical studies demonstrate its anti-tumor activity against tumor cell lines such as SMMC-7721 MCF-7 HCT-116 and NSCLC xenografts underscoring its utility in cancer biology research