Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Camptothecin 97% MP Biomedicals

Catalog No. ICN15973283 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15973283 250 mg
ICN15973225 25 mg
ICN15973250 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15973283 Supplier MP Biomedicals, Inc Supplier No. 0215973283
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3-4-7689
Molecular Formula C20H16N2O4
Molecular Weight (g/mol) 348.36
MDL Number MFCD00081076
InChI Key VSJKWCGYPAHWDS-FQEVSTJZSA-N
Synonym camptothecin, camptothecine, s-+-camptothecin, campathecin, +-camptothecine, d-camptothecin, 20 s-camptothecine, +-camptothecin, 21,22-secocamptothecin-21-oic acid lactone, s-camptothecin
PubChem CID 24360
ChEBI CHEBI:27656
SMILES CCC1(C2=C(COC1=O)C(=O)N3CC4=CC5=CC=CC=C5N=C4C3=C2)O

Specifications

Color Yellow
Density 1.51g/cm3
Assay Percent Range >97%
Quantity 250 mg
Formula Weight 348.3
Physical Form Solid
Chemical Name or Material Camptothecin
Recommended Storage Store at + 4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.