Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Camptothecin, 98%

Catalog No. p-81938
Change view
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC276721000 100 mg Glass bottle
AC276720010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC276721000 Supplier Thermo Scientific Chemicals Supplier No. 276721000
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 7689-03-4 | C20H16N2O4 | 348.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3-4-7689
Molecular Formula C20H16N2O4
Molecular Weight (g/mol) 348.35
MDL Number MFCD00081076
InChI Key VSJKWCGYPAHWDS-FQEVSTJZSA-N
Synonym camptothecin, camptothecine, s-+-camptothecin, campathecin, +-camptothecine, d-camptothecin, 20 s-camptothecine, +-camptothecin, 21,22-secocamptothecin-21-oic acid lactone, s-camptothecin
PubChem CID 24360
ChEBI CHEBI:27656
SMILES CCC1(C2=C(COC1=O)C(=O)N3CC4=CC5=CC=CC=C5N=C4C3=C2)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 260.0°C
Color Yellow
Loss on Drying 1.0% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Merck Index 15, 1737
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,ethanol,acetonirile,dmso,,chloroform,hydrochloric acid
Formula Weight 348.35
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (+)-Camptothecin, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECSNumber UQ0492000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.