Learn More
Medchemexpress LLC Camptothecin-d5 | 1329616-37-6 | 99.6% | 353.38 g/mol | C20H11D5N2O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY16560S10MG
Encompass_Preferred
Camptothecin-d5 is a deuterium-labeled analogue of camptothecin supplied as a solid research standard for analytical and biochemical applications. It is intended for use as an internal standard, tracer, or reference in assays and studies involving camptothecin and related compounds.
  • Deuterium-labeled camptothecin suitable for analytical use.
  • High purity (99.61%) for reliable analytical results.
  • Molecular formula C20H11D5N2O4; molecular weight 353.38 g/mol.
  • Available in small-mass vials (1 mg, 5 mg, 10 mg); larger sizes available by quote.
  • Common applications include internal standardization, ADME studies, and protein-binding or mass-spectrometry assays.
Catalog No. 50-002-89308