Learn More
NEOGEN Campy Blood-Free Selective Medium (Modified CCDA) (EMEA), 500 g
SDP
Catalog No. 502015702
Encompass_Preferred
Campylobacter Blood-Free Selective Medium (Modified Charcoal Cefoperazone Deoycholate Agar or mCCDA) is used for the cultivation of campylobacters, and is not intended for use in the diagnosis of disease or other conditions in humans.A blood free medium which will support the growth of most enteric campylobacters. The selective supplement X112 (X212) makes the medium selective for C. jejuni. and C. coli when incubated at 37°C. With this product incubation at 42°C is no longer necessary and higher recovery rates have been reported at 37°C than at 42°C. The supplement X112 (X212) consists of cefoperazone and amphotericin and is superior to the selective combination of Skirrow, Butzler and Blazer-Wang all of which contain antibiotics shown to be inhibitors to C. coli. The colonial morphologies of Campylobacter spp. on this medium are distinctive.For more information visit https://www.neogen.com/solutions/Microbiology/campy-blood-free-selective-medium-modified-ccda-emea
Catalog No. 50-201-5702 Supplier NEOGEN Supplier No. 700004618
Campylobacter Blood-Free Selective Medium (Modified Charcoal Cefoperazone Deoycholate Agar or mCCDA) is used for the cultivation of campylobacters, and is not intended for use in the diagnosis of disease or other conditions in humans.A blood free medium which will support the growth of most enteric campylobacters. The selective supplement X112 (X212) makes the medium selective for C. jejuni. and C. coli when incubated at 37°C. With this product incubation at 42°C is no longer necessary and higher recovery rates have been reported at 37°C than at 42°C. The supplement X112 (X212) consists of cefoperazone and amphotericin and is superior to the selective combination of Skirrow, Butzler and Blazer-Wang all of which contain antibiotics shown to be inhibitors to C. coli. The colonial morphologies of Campylobacter spp. on this medium are distinctive.For more information visit https://www.neogen.com/solutions/Microbiology/campy-blood-free-selective-medium-modified-ccda-emea