Learn More
NEOGEN Campy Blood-Free Selective Medium (Modified CCDA), 5 kg
SDP
Catalog No. 502015274
Encompass_Preferred
Campy Blood-Free Selective Medium (CCDA) is used with cefoperazone for the selective isolation of Campylobacter spp. in a laboratory setting. Campy Blood-Free Selective Medium (CCDA) is not intended for use in the diagnosis of disease or other conditions in humansCampy Blood-Free Selective Medium (CCDA) was described by Bolton et al. This medium was formulated to replace blood with charcoal, ferrous sulfate, and sodium pyruvate. To improve selectivity, cefoperazone replaced cephazolin in the original formulation. Bolton et al recommended incubating inoculated plates at 37°C to improve isolation rates. Yeast and fungal contaminants are inhibited with the addition of amphotericin B. Campy Blood-Free Selective Medium (CCDA) is recommended for food testing.For more information visit https://www.neogen.com/solutions/Microbiology/campy-blood-free-selective-medium-modified-ccda
Catalog No. 50-201-5274 Supplier NEOGEN Supplier No. 700003070
Campy Blood-Free Selective Medium (CCDA) is used with cefoperazone for the selective isolation of Campylobacter spp. in a laboratory setting. Campy Blood-Free Selective Medium (CCDA) is not intended for use in the diagnosis of disease or other conditions in humansCampy Blood-Free Selective Medium (CCDA) was described by Bolton et al. This medium was formulated to replace blood with charcoal, ferrous sulfate, and sodium pyruvate. To improve selectivity, cefoperazone replaced cephazolin in the original formulation. Bolton et al recommended incubating inoculated plates at 37°C to improve isolation rates. Yeast and fungal contaminants are inhibited with the addition of amphotericin B. Campy Blood-Free Selective Medium (CCDA) is recommended for food testing.For more information visit https://www.neogen.com/solutions/Microbiology/campy-blood-free-selective-medium-modified-ccda