Learn More
NEOGEN Campy Cefex Agar, 10 kg
SDP
Catalog No. 502015440
Encompass_Preferred
Campy Cefex Agar is used for the selective isolation of Campylobacter spp in a laboratory setting. Campy Cefex Agar is not intended for use in the diagnosis of disease or other conditions in humans.Poultry is a primary reservoir of Campylobacter spp. and may cause contamination of more than 80% of chicken carcasses. Campy Cefex Agar is described by Stern et al. This medium was formulated to provide selective isolation of cephalothin-resistant Campylobacter species such as C. jejuni and C. coli.For more information visit https://www.neogen.com/solutions/Microbiology/campy-cefex-agar
Catalog No. 50-201-5440 Supplier NEOGEN Supplier No. 700003249
Campy Cefex Agar is used for the selective isolation of Campylobacter spp in a laboratory setting. Campy Cefex Agar is not intended for use in the diagnosis of disease or other conditions in humans.Poultry is a primary reservoir of Campylobacter spp. and may cause contamination of more than 80% of chicken carcasses. Campy Cefex Agar is described by Stern et al. This medium was formulated to provide selective isolation of cephalothin-resistant Campylobacter species such as C. jejuni and C. coli.For more information visit https://www.neogen.com/solutions/Microbiology/campy-cefex-agar