Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TECHLAB™ CAMPYLOBACTER CHEK™ Diluent

Catalog No. 21279143
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-279-143 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-279-143 Supplier TECHLAB™ Supplier No. 5052DIVD
Add to Cart
Add to Cart

The CAMPYLOBACTER CHEK™ test is an enzyme immunoassay for the qualitative detection of a Campylobacter-specific antigen in human fecal specimens. It is designed to detect C. jejuni, C. coli, C. lari, and C. upsaliensis from patients with signs and symptoms of gastroenteritis.

  • The test is intended for use with preserved fecal specimens in transport media and unpreserved fecal specimens.
  • Test results should be considered in conjunction with clinical findings and patient history.

Specifications

Form Liquid
Format 96-well Plate
Product Type Diluent
Target Organism Class Gram-negative Organisms
Test Type Microbial Detection
Quantity 1 Ea.
Species Campylobacter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.