Learn More
ATCC Campylobacter coli
Supplier:  ATCC 43478
Encompass
Campylobacter coli strain 76-GA2 [LMG 21266] was isolated from marmoset feces. This whole-genome sequenced bacterial strain can be used in media testing, enteric disease research, and emerging infectious disease research.
Catalog No. 50-238-1992