Learn More
NEOGEN Campylobacter Enrichment Broth (Bolton Broth), 5 kg
SDP
Catalog No. 502015430
Encompass_Preferred
Campylobacter Enrichment Broth (Bolton Broth) is used with antimicrobics for the selective enrichment of Campylobacter spp. Campylobacter Enrichment Broth (Bolton Broth) is not intended for use in the diagnosis of disease or other conditions in humans. Campylobacter spp. are microaerophilic, very small, curved, thin, Gram-negative rods. Microaerophilic organisms have a tendency to be more sensitive to toxic forms of oxygen. Campylobacter Enrichment Broth (Bolton), along with nutritional ingredients, contains compounds which enhance the aerotolerance of microaerophilic bacteria by suppressing the toxic form of oxygen. Campylobacter Enrichment Broth (Bolton) is recommended in food testing. Blood-Free Campylobacter Enrichment Broth, Bolton Broth (2X Concentration) is described by the USDA.For more information visit https://www.neogen.com/solutions/Microbiology/campylobacter-enrichment-broth-bolton
Catalog No. 50-201-5430 Supplier NEOGEN Supplier No. 700003231
Campylobacter Enrichment Broth (Bolton Broth) is used with antimicrobics for the selective enrichment of Campylobacter spp. Campylobacter Enrichment Broth (Bolton Broth) is not intended for use in the diagnosis of disease or other conditions in humans. Campylobacter spp. are microaerophilic, very small, curved, thin, Gram-negative rods. Microaerophilic organisms have a tendency to be more sensitive to toxic forms of oxygen. Campylobacter Enrichment Broth (Bolton), along with nutritional ingredients, contains compounds which enhance the aerotolerance of microaerophilic bacteria by suppressing the toxic form of oxygen. Campylobacter Enrichment Broth (Bolton) is recommended in food testing. Blood-Free Campylobacter Enrichment Broth, Bolton Broth (2X Concentration) is described by the USDA.For more information visit https://www.neogen.com/solutions/Microbiology/campylobacter-enrichment-broth-bolton