Learn More
Bioworld CAMPYLOBACTER ENRICHMENT BROTH
Catalog No. 50254291
Encompass_Preferred
Campylobacter Enrichment Broth Used with antimicrobics for the selective enrichment of Campylobacter species; Supplement with Cefoperazone, Cycloheximide, Trimethoprim, and Vancomycin to increase selectivity; May be enriched with Lysed H...
Catalog No. 50-254-291 Supplier Bioworld Supplier No. 306200171
Campylobacter Enrichment Broth Used with antimicrobics for the selective enrichment of Campylobacter species; Supplement with Cefoperazone, Cycloheximide, Trimethoprim, and Vancomycin to increase selectivity; May be enriched with Lysed H...