Learn More
ATCC Campylobacter fetus; subsp. fetus
Catalog No. 502382758
Encompass
Campylobacter fetus subsp. fetus strain [NCTC 10842] was isolated from the brain of a sheep fetus. This whole-genome sequenced bacterial type strain grows best in a biphasic culture.
Catalog No. 50-238-2758 Supplier ATCC Supplier No. 27374
Campylobacter fetus subsp. fetus strain [NCTC 10842] was isolated from the brain of a sheep fetus. This whole-genome sequenced bacterial type strain grows best in a biphasic culture.