Learn More
ATCC Providencia rettgeri; Strain 1103204
Catalog No. 502380481
Encompass
Providencia rettgeri strain bMx# 1103204 is a whole-genome sequenced clinical isolate. The depositor states that this bacterium is a carbapenemase producer (OXA-48).
Catalog No. 50-238-0481 Supplier ATCC Supplier No. BAA2525
Providencia rettgeri strain bMx# 1103204 is a whole-genome sequenced clinical isolate. The depositor states that this bacterium is a carbapenemase producer (OXA-48).