Learn More
Seracare Life Sciences Inc CAMPYLOBACTER GENUS POS CON
Catalog No. 5067463
Encompass_Preferred
Heat-killed Campylobacter jejuni cells. It consists of at least 4.8 x 10^8 cells/mL (1 mL of approximately 2% w/v wet packed cells) in dextran solution. It is intended for use as a positive control in immunoassays designed for Campylobacter detection.
This item is not returnable. View return policy
Catalog No. 50-674-63 Supplier Seracare Life Sciences Inc Supplier No. 53700011
Heat-killed Campylobacter jejuni cells. It consists of at least 4.8 x 10^8 cells/mL (1 mL of approximately 2% w/v wet packed cells) in dextran solution. It is intended for use as a positive control in immunoassays designed for Campylobacter detection.