Learn More
Micro Biologics Inc 3P CAMPYLOBACTER JEJUNI
SDP
Catalog No. NC9537310
Encompass_Preferred
The KWIK-STIK™ is well-known for its clever design and accurate, repeatable results. Each KWIK-STIK™ device features a single QC microorganism strain in a lyophilized pellet, a reservoir of hydrating fluid, and an inoculating swab. For added convenience, a peel-off identification label is included for easy documentation. Campylobacter jejuni subsp. jejuni derived from ATCC® 29428™* 6 self-contained units of a single organism
Catalog No. NC9537310 Supplier Micro Biologics Inc 3P Supplier No. 0325P
The KWIK-STIK™ is well-known for its clever design and accurate, repeatable results. Each KWIK-STIK™ device features a single QC microorganism strain in a lyophilized pellet, a reservoir of hydrating fluid, and an inoculating swab. For added convenience, a peel-off identification label is included for easy documentation. Campylobacter jejuni subsp. jejuni derived from ATCC® 29428™* 6 self-contained units of a single organism