Learn More
Bioss Campylobacter jejuni
Supplier: Bioss BSM72023M
Campylobacter jejuni Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications WB Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide