Learn More
Bioss Campylobacter jejuni
SDP
Supplier:  Bioss BSM72025M
Encompass_Preferred
Campylobacter jejuni Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications WB Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-66440