Learn More
ATCC Campylobacter jejuni; subsp. doylei
Supplier:  ATCC 49349
Encompass
Campylobacter jejuni subsp. doylei strain NCTC 11951 is a bacterial type strain that was isolated in Australia from human feces. This strain has applications in enteric disease research.
Catalog No. 50-238-1085