Learn More
ATCC Campylobacter jejuni; subsp. jejuni
Supplier:  ATCC 33560
Encompass
Campylobacter jejuni subsp. jejuni strain CIP 702 is a whole-genome sequenced bacterial type strain that was isolated from bovine feces. This bacterial strain can be used in enteric and emerging infectious disease research.
Catalog No. 50-238-1934
 Restricted Item