Learn More
ATCC Campylobacter jejuni; subsp. jejuni
Supplier:  ATCC 29428
Encompass
Campylobacter jejuni subsp. jejuni strain VPI H840 was isolated from the feces of a human child. This whole-genome sequenced bacterium is propagated under microaerophilic conditions and can be used in enteric disease research, infectious disease research, and media testing.
Catalog No. 50-238-3344