Learn More
ATCC Crocosphaera subtropica
Catalog No. 502380257
Encompass
Crocosphaera subtropica strain BH68 is a cyanobacterium type strain that was isolated from an intertidal area in Port Aransas in Texas.
Catalog No. 50-238-0257 Supplier ATCC Supplier No. 51142
Crocosphaera subtropica strain BH68 is a cyanobacterium type strain that was isolated from an intertidal area in Port Aransas in Texas.