Learn More
ATCC Borrelia andersonii
Catalog No. 502380066
Encompass
Candidatus Borrelia andersonii strain 21038 is a bacterium that was isolated from tick larvae in Millbrook, New York. This strain is propagated under microaerophilic conditions.
Catalog No. 50-238-0066 Supplier ATCC Supplier No. 700555
Add to Cart
Edge
Add to Cart
Candidatus Borrelia andersonii strain 21038 is a bacterium that was isolated from tick larvae in Millbrook, New York. This strain is propagated under microaerophilic conditions.