Learn More
Aviva Systems Biology Corp Camsap1 Antibody - C-terminal region (ARP58433_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58433_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Camsap1. It was validated on Western Blot. . Alternate names: 9530003A05Rik, C77823, PRO2405, bA100C15.1, RP11-100C15.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-286-475