Learn More
Proteintech Group Inc CAMSAP2 Polyclonal antibody
SDP
Supplier:  Proteintech Group Inc 178801AP150UL
Encompass_Preferred
The CAMSAP2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CAMSAP2. This antibody recognizes human, mouse, rat antigen. The CAMSAP2 antibody has been validated for the following applications: IHC, IP, WB,ELISA analysis.
Catalog No. 50-225-6416