Learn More
Electron Microscopy Sciences Applied Physics LaB6 Cathode for Camscan
SDP
Catalog No. 50949838
Encompass
Lanthanum hexaboride (LaB6) and cerium hexaboride (CeB6) cathodes are ideal for many small spot size applications such as SEM, TEM, surface analysis and metrology, and for high current applications such as microwave tubes, lithography, electron-beam welders, X-ray sources and free electron lasers. Greater brightness from low work function yielding higher currents at lower temps than tungsten.
Type: LaB6
Angle: 90°
Tips: 20 mm
Type: non-shunted
Catalog No. 50-949-838 Supplier Electron Microscopy Sciences Supplier No. 8092220L
Lanthanum hexaboride (LaB6) and cerium hexaboride (CeB6) cathodes are ideal for many small spot size applications such as SEM, TEM, surface analysis and metrology, and for high current applications such as microwave tubes, lithography, electron-beam welders, X-ray sources and free electron lasers. Greater brightness from low work function yielding higher currents at lower temps than tungsten.
Type: LaB6
Angle: 90°
Tips: 20 mm
Type: non-shunted