Learn More
Bellco Glass, Inc. Canister Mounting Plate
SDP
Supplier:  Bellco Glass, Inc. 128811111
Encompass_Preferred
Stabilizes pipette canisters during useStainless steel constructionUse with 38mm od glass pipette canistersDimensions:90 x 90mm. MADE IN THE USA Case qty-6
Catalog No. 50-194-4665