Learn More
Medchemexpress LLC Can508 | 140651-18-9 | 99.6% | 218.22 g/mol | C9H10N6O | 10 MG
SDP
Catalog No. 5000213923
Encompass_Preferred
CAN508 is an ATP-competitive inhibitor selective for the CDK9/cyclin T1 complex, designed for biochemical and cellular research. It has a reported IC50 of 0.35 μM and shows approximately 38-fold selectivity over other CDK/cyclin complexes. The compound is supplied as a high-purity solid and is accompanied by a data sheet, certificate of analysis, and safety data sheet.
  • Potent ATP-competitive inhibition of CDK9/cyclin T1 (IC50 0.35 μM).
  • Approximately 38-fold selectivity versus other CDK/cyclin complexes.
  • High reported purity (99.59%).
  • Supplied as a solid with molecular formula C9H10N6O and molar mass 218.22 g/mol.
  • Provided with data sheet, certificate of analysis, and safety data sheet.
  • Intended for biochemical and cellular research use only.
Catalog No. 50-002-13923 Supplier Medchemexpress LLC Supplier No. HY10042910MG
CAN508 is an ATP-competitive inhibitor selective for the CDK9/cyclin T1 complex, designed for biochemical and cellular research. It has a reported IC50 of 0.35 μM and shows approximately 38-fold selectivity over other CDK/cyclin complexes. The compound is supplied as a high-purity solid and is accompanied by a data sheet, certificate of analysis, and safety data sheet.
  • Potent ATP-competitive inhibition of CDK9/cyclin T1 (IC50 0.35 μM).
  • Approximately 38-fold selectivity versus other CDK/cyclin complexes.
  • High reported purity (99.59%).
  • Supplied as a solid with molecular formula C9H10N6O and molar mass 218.22 g/mol.
  • Provided with data sheet, certificate of analysis, and safety data sheet.
  • Intended for biochemical and cellular research use only.