Learn More
Apexbio Technology LLC Canagliflozin(Synonyms: Invokana, TA-7284, JNJ-28431754, Sulisent, Canagliflozin hemihydrate), 10mM (in 1mL DMSO), CAS: 842133-18-0.
SDP
Catalog No. 5000061053
Encompass_Preferred
Canagliflozin (CAS 842133-18-0) is a selective inhibitor of sodium-glucose cotransporter 2 (SGLT2) a transporter responsible for renal glucose reabsorption By inhibiting SGLT2 activity Canagliflozin decreases renal glucose reabsorption resulting in increased urinary glucose excretion In CHO cells expressing human rat or mouse SGLT2 Canagliflozin potently inhibits sodium-dependent glucose uptake with IC50 values of 4 4 3 7 and 2 0 nM respectively Animal studies using db/db mice and Zucker diabetic fatty (ZDF) rats demonstrate dose-dependent reductions in blood glucose as well as decreased respiratory exchange ratio and body weight Canagliflozin is suitable for in vivo research through oral administration
Catalog No. 50-000-61053 Supplier Apexbio Technology LLC Supplier No. A83335.1
Canagliflozin (CAS 842133-18-0) is a selective inhibitor of sodium-glucose cotransporter 2 (SGLT2) a transporter responsible for renal glucose reabsorption By inhibiting SGLT2 activity Canagliflozin decreases renal glucose reabsorption resulting in increased urinary glucose excretion In CHO cells expressing human rat or mouse SGLT2 Canagliflozin potently inhibits sodium-dependent glucose uptake with IC50 values of 4 4 3 7 and 2 0 nM respectively Animal studies using db/db mice and Zucker diabetic fatty (ZDF) rats demonstrate dose-dependent reductions in blood glucose as well as decreased respiratory exchange ratio and body weight Canagliflozin is suitable for in vivo research through oral administration