Learn More
Selleck Chemical LLC Canagliflozin 5mg 842133-18-0 TA 7284, JNJ 28431754
SDP
Catalog No. 501362917
Encompass_Preferred
Canagliflozin (TA 7284, JNJ 28431754) is a highly potent and selective SGLT2 inhibitor for hSGLT2 with IC50 of 2.2 nM in a cell-free assay, exhibits 413-fold selectivity over hSGLT1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2917 Supplier Selleck Chemical LLC Supplier No. S27605MG
Canagliflozin (TA 7284, JNJ 28431754) is a highly potent and selective SGLT2 inhibitor for hSGLT2 with IC50 of 2.2 nM in a cell-free assay, exhibits 413-fold selectivity over hSGLT1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.