Learn More
Medchemexpress LLC Canagliflozin | 842133-18-0 | MFCD18251436 | 99.7% | 444.52 | C24H25FO5S | 200 MG
SDP
Catalog No. 5000244012
Encompass_Preferred
Canagliflozin is a selective sodium-glucose cotransporter 2 (SGLT2) inhibitor provided as a research-grade powder for biochemical and pharmacological studies. It exhibits low-nanomolar potency against SGLT2 and is used in in vitro and in vivo experiments to investigate glucose transport and metabolic pathways.
  • Selective SGLT2 inhibitor with low-nanomolar IC50 values.
  • High purity suitable for analytical and research use (≈99.7% LCMS).
  • Solid powder form for flexible formulation and dosing.
  • Stable under recommended storage conditions (powder: -20°C and 4°C).
  • Molecular weight 444.52 and formula C24H25FO5S.
Catalog No. 50-002-44012 Supplier Medchemexpress LLC Supplier No. HY10451200MG
Canagliflozin is a selective sodium-glucose cotransporter 2 (SGLT2) inhibitor provided as a research-grade powder for biochemical and pharmacological studies. It exhibits low-nanomolar potency against SGLT2 and is used in in vitro and in vivo experiments to investigate glucose transport and metabolic pathways.
  • Selective SGLT2 inhibitor with low-nanomolar IC50 values.
  • High purity suitable for analytical and research use (≈99.7% LCMS).
  • Solid powder form for flexible formulation and dosing.
  • Stable under recommended storage conditions (powder: -20°C and 4°C).
  • Molecular weight 444.52 and formula C24H25FO5S.