Learn More
Selleck Chemical LLC Canagliflozin hemihydrate
SDP
Catalog No. 5000003544
Encompass_Preferred
Canagliflozin hemihydrate is the hemihydrate form of canagliflozin which is a SGLT2 inhibitor with IC50 of 2 2 nM for hSGLT2 in a cell-free assay exhibits 413-fold selectivity over hSGLT1
Catalog No. 50-000-03544 Supplier Selleck Chemical LLC Supplier No. S590110MG
Canagliflozin hemihydrate is the hemihydrate form of canagliflozin which is a SGLT2 inhibitor with IC50 of 2 2 nM for hSGLT2 in a cell-free assay exhibits 413-fold selectivity over hSGLT1