Learn More
Analytical Sales & Services CANARY-SAFE 10 LITER WASTE
Catalog No. NC2309065
Encompass
Container for B53 and S55 Waste Caps PE-HD190mm x 215mm x 320mm
Catalog No. NC2309065 Supplier Analytical Sales & Services Supplier No. CL53010
Container for B53 and S55 Waste Caps PE-HD190mm x 215mm x 320mm