Learn More
Analytical Sales & Services Canary-Safe Air Check Valve, Prep Size
Catalog No. 5000805373
Encompass
Canary-Safe PREP Size Air Check Valve for Mobile Phase Safety Caps\n\nSuggested for Flow Rates >100mL and <500mL
Catalog No. 50-008-05373 Supplier Analytical Sales & Services Supplier No. CX0002
Canary-Safe PREP Size Air Check Valve for Mobile Phase Safety Caps\n\nSuggested for Flow Rates >100mL and <500mL