Learn More
Analytical Sales & Services Canary Safe B53 Mobile Phase Cap, 4 Ports for 3/16" Tubing
Catalog No. 5000805287
Encompass
Canary Safe B53 Mobile Phase Cap, 4 Ports for 3/16" Tubing
Catalog No. 50-008-05287 Supplier Analytical Sales & Services Supplier No. CC53416
Add to Cart
Add to Cart
Canary Safe B53 Mobile Phase Cap, 4 Ports for 3/16" Tubing