Learn More
Analytical Sales & Services Canary-Safe Cap, GL38, 1 Port for 3/16" Tubing
Catalog No. 5000805330
Encompass
Canary-Safe Cap, GL38\n\n• 1 Port for 3/16" Tubing\n• Air Check Valve Port\n• Includes fittings and Air Check Valve\n\nQty/Pkg: EACH
Catalog No. 50-008-05330 Supplier Analytical Sales & Services Supplier No. CCGL381316
Canary-Safe Cap, GL38\n\n• 1 Port for 3/16" Tubing\n• Air Check Valve Port\n• Includes fittings and Air Check Valve\n\nQty/Pkg: EACH