Learn More
Analytical Sales & Services Canary-Safe Cap, GL38, 1 Standard Port, 1 Port for 3/16" Tubing
Catalog No. 5000807102
Encompass
Canary-Safe Cap, GL38\n\n• 1 Standard (1/8") Tubing Port\n• 1 Port for 3/16" Tubing\n• Air Check Valve Port\n• Includes fittings and Air Check Valve\n\nQty/Pkg: EACH
Catalog No. 50-008-07102 Supplier Analytical Sales & Services Supplier No. CCGL381836
Canary-Safe Cap, GL38\n\n• 1 Standard (1/8") Tubing Port\n• 1 Port for 3/16" Tubing\n• Air Check Valve Port\n• Includes fittings and Air Check Valve\n\nQty/Pkg: EACH