Learn More
Analytical Sales & Services Canary-Safe Cap GL38, Mobile Phase Bottle Safety Cap, Prevents Solvent Evaporation, 3 Standard Ports, Unit: each
Catalog No. NC1812769
Encompass
The Canary-Safe Cap GL38 with 3 Standard Tubing Ports is designed for secure solvent management, enhancing laboratory safety by preventing evaporation. With its GL38 thread compatibility and no need for adapters, it provides seamless integration with 4L amber bottles. This cap features a polypropylene shell and PTFE core, along with essential fittings and an air check valve, making it a reliable choice for scientific environments.
Catalog No. NC1812769 Supplier Analytical Sales & Services Supplier No. CCGL3803
The Canary-Safe Cap GL38 with 3 Standard Tubing Ports is designed for secure solvent management, enhancing laboratory safety by preventing evaporation. With its GL38 thread compatibility and no need for adapters, it provides seamless integration with 4L amber bottles. This cap features a polypropylene shell and PTFE core, along with essential fittings and an air check valve, making it a reliable choice for scientific environments.