Learn More
Analytical Sales & Services Canary-Safe Cap, GL45, 1 Standard Port and 1 Port for 3/16" Tubing
Catalog No. 5000805269
Encompass
Canary-Safe Cap, GL45 \n\n• 1 Standard (1/8") Tubing Port\n• 1 Port for 3/16" Tubing\n• Air Check Valve Port\n• Includes fittings and Air Check Valve\n\nQty/Pkg: Each
Catalog No. 50-008-05269 Supplier Analytical Sales & Services Supplier No. CC2316
Canary-Safe Cap, GL45 \n\n• 1 Standard (1/8") Tubing Port\n• 1 Port for 3/16" Tubing\n• Air Check Valve Port\n• Includes fittings and Air Check Valve\n\nQty/Pkg: Each