Learn More
Analytical Sales & Services Canary-Safe Cap, GL45, 2 Ports for 3/16" Tubing
Catalog No. 5000805333
Encompass
Canary-Safe Cap, GL45 \n• 2 Port for 3/16" Tubing\n• Air Check Valve Port\n• Includes fittings and Air Check Valve\n\nQty/Pkg: Each\n\n \n\nCC2236 - $176; Canary-Safe Cap (GL45), 2 Ports for 3/16" Tubing, 1 Port for Air Check Valve
Catalog No. 50-008-05333 Supplier Analytical Sales & Services Supplier No. CC2236
Canary-Safe Cap, GL45 \n• 2 Port for 3/16" Tubing\n• Air Check Valve Port\n• Includes fittings and Air Check Valve\n\nQty/Pkg: Each\n\n \n\nCC2236 - $176; Canary-Safe Cap (GL45), 2 Ports for 3/16" Tubing, 1 Port for Air Check Valve