Learn More
Analytical Sales & Services Canary-Safe Cap, GL45, 4 Ports for 3/16" Tubing
Catalog No. 5000805270
Encompass
Canary-Safe Mobile Phase Cap, GL45 \n\n• 4 Ports for 3/16" Tubing\n• Air Check Valve port\n• Includes fittings and Air Check Valve\n\nQty/Pkg: Each
Catalog No. 50-008-05270 Supplier Analytical Sales & Services Supplier No. CC4316
Canary-Safe Mobile Phase Cap, GL45 \n\n• 4 Ports for 3/16" Tubing\n• Air Check Valve port\n• Includes fittings and Air Check Valve\n\nQty/Pkg: Each