Learn More
Analytical Sales & Services Canary-Safe Cap I, GL45, 1 Standard Tubing Port with Shut-Off Valve
Catalog No. 5000805226
Encompass
Canary-Safe Cap I, GL45, with Shut-Off Valve, 1 Standard Port for 1/8" Tubing\n\nQty/Pkg: EACH
Catalog No. 50-008-05226 Supplier Analytical Sales & Services Supplier No. CC1011
Canary-Safe Cap I, GL45, with Shut-Off Valve, 1 Standard Port for 1/8" Tubing\n\nQty/Pkg: EACH