Learn More
Analytical Sales & Services Canary-Safe Cap, S55, with 8 Ports for Standard Tubing
Catalog No. 5000805288
Encompass
Canary-Safe S55 Mobile Phase Cap with 8 Ports for Standard 1/8" Tubing\n\nIncludes Filter Check Valve and fittings
Catalog No. 50-008-05288 Supplier Analytical Sales & Services Supplier No. CC5508
Canary-Safe S55 Mobile Phase Cap with 8 Ports for Standard 1/8" Tubing\n\nIncludes Filter Check Valve and fittings