Learn More
Analytical Sales & Services Canary-Safe DARK BLUE Cap 5, GL45, 5 Standard Tubing Ports
Catalog No. 5000805382
Encompass
Canary-Safe™ Cap 5\n\n• GL45 Threads\n• 5 Standard Ports for 1/8" Tubing\n• Includes Air Check Valve\n• Color: Dark Blue\n\nQty/Pkg: EACH
Catalog No. 50-008-05382 Supplier Analytical Sales & Services Supplier No. CC1005B
Add to Cart
Add to Cart
Canary-Safe™ Cap 5\n\n• GL45 Threads\n• 5 Standard Ports for 1/8" Tubing\n• Includes Air Check Valve\n• Color: Dark Blue\n\nQty/Pkg: EACH