Learn More
Analytical Sales & Services Canary-Safe Waste Cap, B53, 1 Standard Tubing Port, 3 Ports for Large ID Tubing
Catalog No. 5000805370
Encompass
Canary-Safe Waste Cap, B53\n\n• 1 Standard (1/8") Tubing Port\n• 3 Ports for Large ID Tubing\n• Charcoal Exhaust Filter Port\n• Includes fittings (Charcoal Exhaust Filter sold separately)\n\nQty/Pkg: Each
Catalog No. 50-008-05370 Supplier Analytical Sales & Services Supplier No. CW53103
Canary-Safe Waste Cap, B53\n\n• 1 Standard (1/8") Tubing Port\n• 3 Ports for Large ID Tubing\n• Charcoal Exhaust Filter Port\n• Includes fittings (Charcoal Exhaust Filter sold separately)\n\nQty/Pkg: Each