Learn More
Analytical Sales & Services Canary-Safe Waste Cap, B53, with 2 Standard Tubing Ports
Catalog No. 5000807145
Encompass
Canary-Safe Waste Cap B53 for Nalgene 2 Ports for 1/8" OD Tubing 1 Port for Charcoal Exhaust Filter
Catalog No. 50-008-07145 Supplier Analytical Sales & Services Supplier No. CW53219
Add to Cart
Add to Cart
Canary-Safe Waste Cap B53 for Nalgene 2 Ports for 1/8" OD Tubing 1 Port for Charcoal Exhaust Filter