Learn More
Analytical Sales & Services Canary-Safe Waste Cap, B53, with 4 Standard Tubing Ports & 1 Port for Barbed Adapter
Catalog No. 5000805139
Encompass
Canary-Safe Waste Cap B53 for Nalgene 4 Ports for 1/8" OD Tubing 1 Port for Barbed Adapter 1 Port for Charcoal Exhaust Filter
Catalog No. 50-008-05139 Supplier Analytical Sales & Services Supplier No. CW53418
Canary-Safe Waste Cap B53 for Nalgene 4 Ports for 1/8" OD Tubing 1 Port for Barbed Adapter 1 Port for Charcoal Exhaust Filter