Learn More
Analytical Sales & Services Canary-Safe Waste Cap, B53, with Safety Funnel, 1 Standard Port & 1 Port for Barbed Adapter
Catalog No. 5000805351
Encompass
Canary-Safe Waste Cap 53B with Safety Funnel 1 Standard Port 1 Port for Barbed Adapter 1 Exhaust Filter Port
Catalog No. 50-008-05351 Supplier Analytical Sales & Services Supplier No. CSF5318
Canary-Safe Waste Cap 53B with Safety Funnel 1 Standard Port 1 Port for Barbed Adapter 1 Exhaust Filter Port