Learn More
Analytical Sales & Services Canary-Safe Waste Cap, B83, with 1 Standard Tubing Port & 4 Ports for Barbed Adapters
Catalog No. 5000805371
Encompass
Canary-Safe Waste Cap\n\n• Thread size: B83\n• Standard Tubing Ports: 1\n• Ports for Barbed Adapters: 4\n• Port for Charcoal Exhaust Filter
Catalog No. 50-008-05371 Supplier Analytical Sales & Services Supplier No. CW83401
Canary-Safe Waste Cap\n\n• Thread size: B83\n• Standard Tubing Ports: 1\n• Ports for Barbed Adapters: 4\n• Port for Charcoal Exhaust Filter