Learn More
Analytical Sales & Services Canary-Safe Waste Cap, B83, with 2 Standard Tubing Ports & 3 Ports for Barbed Adapters
Catalog No. 5000805296
Encompass
Canary-Safe Waste Cap\n\n• Thread size: B83\n• Standard Tubing Ports: 2\n• Ports for Barbed Adapters: 3\n• Port for Charcoal Exhaust Filter
Catalog No. 50-008-05296 Supplier Analytical Sales & Services Supplier No. CW83302
Canary-Safe Waste Cap\n\n• Thread size: B83\n• Standard Tubing Ports: 2\n• Ports for Barbed Adapters: 3\n• Port for Charcoal Exhaust Filter